Pentylbenzene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenylpentane, Amylbenzene

A15734-22 A15734-06 A15734-14
AAA15734-22EA 214 USD
AAA15734-22 AAAA15734-06 AAA15734-14
Pentylbenzene 96%
Pentylbenzene

Purity: 96% CAS No.: 538-68-1 Molecular Formula: C11H16 Formula Weight: 148.25 Flash Point: 65°(149°F) Refractive Index: 1.4880 Melting Point: -75° Boiling Point: 204-205° Beilstein Reference: 1904359 Merck Reference: 14,600 UN No.: 3082 MDL No.: MFCD00009502

Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: -1 °C
Density: 0.8594 g/cm³ (20 °C)
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009502
CAS Number: 538-68-1
EINECS: 208-701-5
UN: 3082
ADR: 9,III
Merck Index: 13,00604

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR