Dicyclohexylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DCHA

A15671-AE A15671-AP
AAAA15671-AEEA 34.8 USD
AAAA15671-AE AAA15671-AP
Dicyclohexylamine 98%
Dicyclohexylamine

Purity: 98% CAS No.: 101-83-7 Molecular Formula: C12H23N Formula Weight: 181.32 Flash Point: 99°(210°F) Refractive Index: 1.4842 Melting Point: -2° Boiling Point: 256° Beilstein Reference: 605923 Merck Reference: 14,3095 UN No.: 2565 MDL No.: MFCD00011658

Formula: C₁₂H₂₃N
MW: 181.32 g/mol
Boiling Pt: 256 °C
Melting Pt: –2 °C
Density: 0.91 g/cm³ (20 °C)
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011658
CAS Number: 101-83-7
EINECS: 202-980-7
UN: 2565
ADR: 8,III
Merck Index: 13,03122

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR