2-Bromo-1,4-dimethoxybenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2,5-dimethoxybenzene

A15640-09 A15640-18 A15640-30
AAA15640-09EA 61.5 USD
AAA15640-09 AAA15640-18 AAA15640-30
2-Bromo-1,4-dimethoxybenzene 98%
2-Bromo-1,4-dimethoxybenzene

Purity: 98% CAS No.: 25245-34-5 Molecular Formula: C8H9BrO2 Formula Weight: 217.07 Flash Point: 110°(230°F) Refractive Index: 1.5700 Boiling Point: 260-262° Beilstein Reference: 2441884 MDL No.: MFCD00008355

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 107 °C (0.2 -rr)
Density: 1.445 g/cm³ (15 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR