Cyclohexyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

A15585-14 A15585-06
AAA15585-14EA 50.61 USD
AAA15585-14 AAAA15585-06
Cyclohexyl isothiocyanate 98%
Cyclohexyl isothiocyanate
Formula: C₇H₁₁NS
Boiling Pt: 219 °C
Melting Pt: 70 - 72 °C
Density: 1.0339 g/cm³ (20 °C)
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003841
CAS Number: 1122-82-3
EINECS: 214-361-9
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR