4-Methylthiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15428-06
A15428-22
A15428-14
AAA15428-06EA
25.9
USD
AAA15428-06
AAAA15428-22
AAA15428-14
4-Methylthiazole 99%
4-Methylthiazole
Purity: 99%
CAS No.: 693-95-8
Molecular Formula: C4H5NS
Formula Weight: 99.16
Flash Point: 32°(89°F)
Refractive Index: 1.5240
Odor: Characteristic
Boiling Point: 133-134°
Beilstein Reference: 105228
UN No.: 1993
MDL No.: MFCD00005340
Formula:
C₄H₅NS MW: 99.16 g/mol Boiling Pt: 133…134 °C Density: 1.1187 g/cm³ (20 °C) Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005340 CAS Number: 693-95-8 EINECS: 211-764-1 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...