Benzotriazole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

A15423-18 A15423-0B A15423-30
AAA15423-18EA 40.9 USD
AAA15423-18 AAA15423-0B AAA15423-30
Benzotriazole 99%
Benzotriazole

Purity: 99% CAS No.: 95-14-7 Molecular Formula: C6H5N3 Formula Weight: 119.13 Flash Point: 212°(413°F) Physical Form: Powder Density: 1.360 Appearance: White to pale cream Odor: Odorless pH: 6.0-7.0 Melting Point: 96-99° Boiling Point: 204°/15mm Beilstein Reference: 112133 Merck Reference: 14,1108 MDL No.: MFCD00005699

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 204 °C/15 mm
Melting Pt: 96…99 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 212 °C (413 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1
UN: 3077
ADR: 9,III
Merck Index: 13,01109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR