1-Chloro-4-nitrobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 98+%
CAS No.: 100-00-5
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Flash Point: 127°(260°F)
Melting Point: 82-86°
Boiling Point: 238-242°
Beilstein Reference: 508691
Merck Reference: 14,2151
UN No.: 1578
MDL No.: MFCD00007285
Formula:
C₆H₄ClNO₂ Boiling Pt: 238…242 °C Melting Pt: 82…86 °C Density: 1.515 Flash Pt: 127 °C (260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007285 CAS Number: 100-00-5 EINECS: 202-809-6 UN: 1578 ADR: 6.1,II Merck Index: 14,2151 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...