2-Chloro-4-fluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A15345-03 A15345-14 A15345-06
AAA15345-03EA 15.8 USD
AAA15345-03 AAAA15345-14 AAA15345-06
2-Chloro-4-fluorobenzaldehyde 97%
2-Chloro-4-fluorobenzaldehyde

Purity: 97% CAS No.: 84194-36-5 Molecular Formula: C7H4ClFO Formula Weight: 158.56 Flash Point: 110°(230°F) Melting Point: 60-63° Boiling Point: 118-120°/50mm Beilstein Reference: 3537704 MDL No.: MFCD00042527

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 118…120 °C/50 mm
Melting Pt: 60…63 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042527
CAS Number: 84194-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR