Methyl-3-hydroxy-4-nitrobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-hydroxy-4-nitrobenzoate

A15329-06 A15329-14
AAA15329-06EA 84.7 USD
AAA15329-06 AAA15329-14 AAAA15329-22
Methyl-3-hydroxy-4-nitrobenzoate 98%
Methyl-3-hydroxy-4-nitrobenzoate

Purity: 98% CAS No.: 713-52-0 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 89-92° Beilstein Reference: 2583832

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00016997
CAS Number: 713-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR