5-Iodovanillin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3-iodo-5-methoxybenzaldehyde

A15162-30 A15162-09 A15162-18
AAA15162-30EA 784 USD
AAA15162-30 AAA15162-09 AAA15162-18
5-Iodovanillin 98%
5-Iodovanillin

Purity: 98% CAS No.: 5438-36-8 Molecular Formula: C8H7IO3 Formula Weight: 278.05 Odor: Irritating Melting Point: 181-184° Beilstein Reference: 2049762 MDL No.: MFCD00006941

Formula: C₈H₇IO₃
MW: 278.05 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006941
CAS Number: 5438-36-8
EINECS: 226-617-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR