2,4-Dichloropyrimidine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15131-30
A15131-06
A15131-18
A15131-09
AAAA15131-30EA
1185
USD
AAAA15131-30
AAAA15131-06
AAA15131-18
AAA15131-09
2,4-Dichloropyrimidine 98+%
2,4-Dichloropyrimidine
Purity: 98+%
CAS No.: 3934-20-1
Molecular Formula: C4H2Cl2N2
Formula Weight: 148.98
Storage Temperature: 2°C to 8°C
Melting Point: 57-62°
Boiling Point: 101°/23mm
Beilstein Reference: 110911
MDL No.: MFCD00006061
Formula:
C₄H₂Cl₂N₂ Boiling Pt: 101 °C/23 mm Melting Pt: 57…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006061 CAS Number: 3934-20-1 EINECS: 223-508-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...