4-Hydroxyphthalic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15091-06
A15091-22
A15091-14
AAA15091-06EA
122
USD
AAA15091-06
AAA15091-22
AAA15091-14
4-Hydroxyphthalic acid 98%
4-Hydroxyphthalic acid
Purity: 98%
CAS No.: 610-35-5
Molecular Formula: C8H6O5
Formula Weight: 182.13
Melting Point: ca 190° dec.
MDL No.: MFCD00013984
Formula:
C₈H₆O₅ MW: 182.13 g/mol Boiling Pt: 386 °C (1013 hPa) Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013984 CAS Number: 610-35-5 EINECS: 210-221-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...