3,5-Dimethoxyaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

A15073-09 A15073-14 A15073-18
AAA15073-09EA 62.2 USD
AAA15073-09 AAAA15073-14 AAA15073-18
3,5-Dimethoxyaniline 98%
3,5-Dimethoxyaniline

Purity: 98% CAS No.: 10272-07-8 Molecular Formula: C8H11NO2 Formula Weight: 153.18 Flash Point: 110°(230°F) Odor: Characteristic Boiling Point: 178°/20mm Beilstein Reference: 638013 UN No.: 2811 MDL No.: MFCD00008392

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 178 °C/20 mm
Melting Pt: 53…57 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR