2-Bromo-5-fluorophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-4-fluoro-2-hydroxybenzene
A15064-06
A15064-14
AAA15064-06EA
43.8
USD
AAA15064-06
AAAA15064-14
AAAA15064-22
2-Bromo-5-fluorophenol 97%
2-Bromo-5-fluorophenol
Purity: 97%
CAS No.: 147460-41-1
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Flash Point: 87°(188°F)
Refractive Index: 1.5540
Boiling Point: 184-186°
Beilstein Reference: 7915214
MDL No.: MFCD00040939
Formula:
C₆H₄BrFO Boiling Pt: 184 - 186 °C (1013 hPa) Density: 1.717 g/cm³ (20 °C) Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040939 CAS Number: 147460-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...