Malononitrile 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15046-0E
A15046-22
A15046-36
AAA15046-0EEA
502
USD
AAA15046-0E
AAA15046-22
AAA15046-36
Malononitrile 99%
Malononitrile
Crosslinking agent for modification of proteins via amidation
Purity: 99%
CAS No.: 109-77-3
Molecular Formula: C3H2N2
Formula Weight: 66.06
Flash Point: 112°(233°F)
Refractive Index: 1.4150
Melting Point: 30-34°
Boiling Point: 220-222°
Solubility: Soluble in water
Beilstein Reference: 773697
Merck Reference: 14,5711
UN No.: 2647
MDL No.: MFCD00001883
Formula:
C₃H₂N₂ MW: 66.06 g/mol Boiling Pt: 220 °C (1013 hPa) Melting Pt: 30 - 32 °C Density: 1.191 g/cm³ (20 °C) Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001883 CAS Number: 109-77-3 EINECS: 203-703-2 UN: 2647 ADR: 6.1,II Merck Index: 13,05733 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...