2-Pyridylmethanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Pyridylmethanol, 2-Pyridylcarbinol, 2-Pyridinemethanol, Pyridin-2-ylmethanol, 2-(Hydroxymethyl)pyridine, Pyridine-2-methanol
A15005-22
A15005-36
A15005-14
AAA15005-22EA
124
USD
AAA15005-22
AAA15005-36
AAA15005-14
2-Pyridylmethanol 98+%
2-Pyridylmethanol
Purity: 98+%
CAS No.: 586-98-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Storage Temperature: 2°C to 8°C
Flash Point: 120°(248°F)
Refractive Index: 1.5440
Melting Point: -6 to -5°
Boiling Point: 112-113°/16mm
Beilstein Reference: 107849
MDL No.: MFCD00006348
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 223 °C (1013 hPa) Melting Pt: -6 °C Density: 1.1317 g/cm³ (20 °C) Flash Pt: 120 °C (248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006348 CAS Number: 586-98-1 EINECS: 209-592-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...