2-Chloro-4-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14953-14 A14953-06
AAA14953-14EA 84 USD
AAA14953-14 AAA14953-06
2-Chloro-4-fluorobenzoic acid 98%
2-Chloro-4-fluorobenzoic acid

Purity: 98% CAS No.: 2252-51-9 Molecular Formula: C7H4ClFO2 Formula Weight: 174.56 Melting Point: 183-187° Beilstein Reference: 1946215

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00010615
CAS Number: 2252-51-9
EINECS: 218-845-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR