Acenaphthene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dihydroacenaphthylene

A14922-0B A14922-30
AAA14922-0BEA 269 USD
AAA14922-0B AAA14922-30
Acenaphthene 97%
Acenaphthene

Purity: 97% CAS No.: 83-32-9 Molecular Formula: C12H10 Formula Weight: 154.21 Flash Point: 135°(275°F) Melting Point: 92-95° Boiling Point: 278° Beilstein Reference: 386081 Merck Reference: 14,28 UN No.: 3077 MDL No.: MFCD00003807

Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 90 - 93 °C
Density: 1.024 g/cm³ (20 °C)
Flash Pt: 135 °C (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00003807
CAS Number: 83-32-9
EINECS: 201-469-6
UN: 3077
ADR: 9,III
Merck Index: 13,00029
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5520bcac8d97ebc91a4b9c3873c734cd4abd6427&vid=0cd04594d640995040d469a56bffa409d582f3ce&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR