4-Cyanobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Formylbenzonitrile

A14914-14 A14914-22 A14914-06
AAA14914-14EA 217.25 USD
AAA14914-14 AAA14914-22 AAA14914-06
4-Cyanobenzaldehyde 98+%
4-Cyanobenzaldehyde

Purity: 98+% CAS No.: 105-07-7 Molecular Formula: C8H5NO Formula Weight: 131.13 Odor: Characteristic Melting Point: 99-103° Boiling Point: 133°/12mm Beilstein Reference: 606473 MDL No.: MFCD00003376

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 133 °C (12 mmHg)
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003376
CAS Number: 105-07-7
EINECS: 203-267-3
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR