4-Bromophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

A14873-22 A14873-36 A14873-0E
AAA14873-22EA 88.62 USD
AAA14873-22 AAA14873-36 AAA14873-0E
4-Bromophenol 99%
4-Bromophenol

Purity: 99% CAS No.: 106-41-2 Molecular Formula: C6H5BrO Formula Weight: 173.01 Flash Point: 110°(230°F) Physical Form: Cryst powder Density: 1.840 Appearance: White to pale brown Odor: Phenol-like Melting Point: 62-66° Boiling Point: 235-238° Beilstein Reference: 1680024 Merck Reference: 14,1428 UN No.: 2811 MDL No.: MFCD00002313

Formula: C₆H₅BrO
Boiling Pt: 235…238 °C
Melting Pt: 62…66 °C
Density: 1.840
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002313
CAS Number: 106-41-2
EINECS: 203-394-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,1428

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR