2-Hexanone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl butyl ketone, Butyl methyl ketone
A14867-22
A14867-36
A14867-09
A14867-18
AAA14867-22EA
185
USD
AAA14867-22
AAA14867-36
AAAA14867-09
AAAA14867-18
2-Hexanone 98%
2-Hexanone
Purity: 98%
CAS No.: 591-78-6
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 23°(73°F)
Refractive Index: 1.4010
Odor: Pungent
Melting Point: -56°
Boiling Point: 126-127°
Beilstein Reference: 1737676
Merck Reference: 14,6033
UN No.: 1224
MDL No.: MFCD00009482
Formula:
C₆H₁₂O Boiling Pt: 126…127 °C Melting Pt: –56 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 23 °C(73 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009482 CAS Number: 591-78-6 EINECS: 209-731-1 UN: 1224 ADR: "3",III Merck Index: 13,06060 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...