Hydroquinone monomethyl ether 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14845-18
A14845-0B
A14845-30
AAA14845-18EA
23.87
USD
AAA14845-18
AAA14845-0B
AAA14845-30
Hydroquinone monomethyl ether 98+%
Hydroquinone monomethyl ether
Purity: 98+%
CAS No.: 150-76-5
Molecular Formula: C7H8O2
Formula Weight: 124.14
Flash Point: 132°(269°F)
Melting Point: 55-58°
Boiling Point: 243-246°
Beilstein Reference: 507924
MDL No.: MFCD00002332
Formula:
C₇H₈O₂ Boiling Pt: 243…246 °C Melting Pt: 55…58 °C Density: 1.550 Flash Pt: 132 °C(269 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002332 CAS Number: 150-76-5 EINECS: 205-769-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...