Isophorone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5,5-Trimethyl-2-cyclohexen-1-one, 3,5,5-Trimethylcyclohex-2-enone
A14844-AK
A14844-0F
A14844-AP
AAAA14844-AKEA
26.5
USD
AAAA14844-AK
AAA14844-0F
AAA14844-AP
Isophorone 97%
Isophorone
Purity: 97%
CAS No.: 78-59-1
Molecular Formula: C9H14O
Formula Weight: 138.21
Flash Point: 84°(183°F)
Refractive Index: 1.4780
Melting Point: -8°
Boiling Point: 99°/18mm
Beilstein Reference: 1280721
Merck Reference: 14,5196
MDL No.: MFCD00001584
Formula:
C₉H₁₄O MW: 138.21 g/mol Boiling Pt: 99 °C/18 mm Melting Pt: –8 °C Density: 0.922 g/cm³ (20 °C) Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001584 CAS Number: 78-59-1 EINECS: 201-126-0 Merck Index: 13,05215 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...