2-Chloro-5-nitrobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A14824-09 A14824-18
AAAA14824-22EA 275.01 USD
AAAA14824-22 AAA14824-09 AAA14824-18
2-Chloro-5-nitrobenzaldehyde 97%
2-Chloro-5-nitrobenzaldehyde

Purity: 97% CAS No.: 6361-21-3 Molecular Formula: C7H4ClNO3 Formula Weight: 185.57 Melting Point: 77-81° Beilstein Reference: 743764

Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS Number: 6361-21-3
EINECS: 228-830-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR