3,6-Dichloropyridazine 98%

Supplier: Thermo Scientific Chemicals
Danger

A14795-22 A14795-14
AAA14795-22EA 95.1 USD
AAA14795-22 AAA14795-36 AAA14795-14
3,6-Dichloropyridazine 98%
3,6-Dichloropyridazine

Purity: 98% CAS No.: 141-30-0 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Melting Point: 66-70° Beilstein Reference: 111151 UN No.: 2811 MDL No.: MFCD00006466

Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 89 - 91 °C (0.2 -rr)
Melting Pt: 66…70 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR