4-Fluorostyrene 97+% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Fluoro-4-vinylbenzene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.1%
A14775-14
A14775-06
A14775-22
A14775-03
AAA14775-14EA
313
USD
AAA14775-14
AAA14775-06
AAA14775-22
AAAA14775-03
4-Fluorostyrene 97+% stabilized
4-Fluorostyrene
Purity: 97+%
CAS No.: 405-99-2
Molecular Formula: C8H7F
Formula Weight: 122.14
Storage Temperature: 2°C to 8°C
Flash Point: 26°(78°F)
Refractive Index: 1.5150
Odor: Recognizable
Melting Point: -36°
Boiling Point: 145-148°
Beilstein Reference: 1634203
UN No.: 1993
MDL No.: MFCD00000361
Formula:
C₈H₇F MW: 122.14 g/mol Boiling Pt: 145…148 °C Melting Pt: –36 °C Density: 1.022 g/cm³ (20 °C) Flash Pt: 26 °C (78 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000361 CAS Number: 405-99-2 EINECS: 206-975-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...