(R)-(-)-Mandelic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid

A14767-14 A14767-22
AAA14767-14EA 72.1 USD
AAA14767-14 AAA14767-06 AAA14767-22
(R)-(-)-Mandelic acid 98%
(R)-(-)-Mandelic acid

Purity: 98% CAS No.: 611-71-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 130-134° Beilstein Reference: 2691094

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 130 - 132 °C
Density: 1.341 g/cm³ (20 °C)
Flash Pt: ≥ 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS Number: 611-71-2
EINECS: 210-276-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR