4-Bromobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A14755-14 A14755-22 A14755-30
AAAA14755-14EA 47.7 USD
AAAA14755-14 AAAA14755-22 AAA14755-30
4-Bromobenzophenone 98%
4-Bromobenzophenone

Purity: 98% CAS No.: 90-90-4 Molecular Formula: C13H9BrO Formula Weight: 261.12 Odor: Odorless Melting Point: 79-83° Boiling Point: 350° Beilstein Reference: 1910182 MDL No.: MFCD00000103

Formula: C₁₃H₉BrO
Boiling Pt: 350 °C
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000103
CAS Number: 90-90-4
EINECS: 202-024-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR