Cytosine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14731-06 A14731-14 A14731-22
AAA14731-06EA 97.87 USD
AAA14731-06 AAA14731-14 AAA14731-22
Cytosine 98+%
Cytosine

Purity: 98+% CAS No.: 71-30-7 Molecular Formula: C4H5N3O Formula Weight: 111.10 Physical Form: Powder Appearance: White Melting Point: 300° Beilstein Reference: 2637 Merck Reference: 14,2795 MDL No.: MFCD00006034

Formula: C₄H₅N₃O
Melting Pt: 313 - 316 °C
Density: 0.99771 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR