2'-Chloroacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Chlorophenyl)acetamide

A14711-36 A14711-14 A14711-22
AAA14711-36EA 244 USD
AAA14711-36 AAA14711-14 AAA14711-22
2'-Chloroacetanilide 98+%
2'-Chloroacetanilide

Purity: 98+% CAS No.: 533-17-5 Molecular Formula: C8H8ClNO Formula Weight: 169.61 Melting Point: 86-88° Beilstein Reference: 2086967 Merck Reference: 14,2111 MDL No.: MFCD00045169

Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00045169
CAS Number: 533-17-5
EINECS: 208-555-2
Merck Index: 13,02128

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR