trans-Cinnamaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14689-0E A14689-22 A14689-36
AAA14689-0EEA 164 USD
AAA14689-0E AAA14689-22 AAA14689-36
trans-Cinnamaldehyde 98+%
trans-Cinnamaldehyde

Purity: 98+% CAS No.: 14371-10-9 Molecular Formula: C9H8O Formula Weight: 132.16 Flash Point: 131°(268°F) Physical Form: Liquid Density: 1.050 Refractive Index: 1.6220 Sensitivity: Air Sensitive Appearance: Yellow Odor: Characteristic Melting Point: -8° Boiling Point: 250-252° Beilstein Reference: 1071571 Merck Reference: 14,2297 MDL No.: MFCD00007000

Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 250…252 °C
Melting Pt: –8 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 131 °C(268 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007000
CAS Number: 14371-10-9
Merck Index: 12,02319

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR