Methyl-3,4-diaminobenzoate 98%

Supplier: Thermo Scientific Chemicals
Danger

A14656-14 A14656-06
AAA14656-14EA 328 USD
AAA14656-14 AAA14656-06
Methyl-3,4-diaminobenzoate 98%
Methyl-3,4-diaminobenzoate

Purity: 98% CAS No.: 36692-49-6 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 105-108° Beilstein Reference: 2091675

Formula: C₈H₁₀N₂O₂
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00017098
CAS Number: 36692-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR