Sodium-1-decanesulphonate, (max. 1,5% H₂O) 99% (dry weight)
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt
A14638-14
A14638-22
A14638-06
AAA14638-14EA
253
USD
AAA14638-14
AAA14638-22
AAA14638-06
Sodium-1-decanesulphonate, (max. 1,5% H₂O) 99% (dry weight)
Sodium-1-decanesulphonate
Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.
Purity: 99%
CAS No.: 13419-61-9
Molecular Formula: CH3(CH2)9SO3Na
Formula Weight: 244.33
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point:
Ion-associating reagent for HPLC, useful for analysis of peptides and proteins.
Formula:
C₁₀H₂₁NaO₃S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007526 CAS Number: 13419-61-9 EINECS: 236-525-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...