4'-Phenylacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Acetylbiphenyl, 1-([1,1'-Biphenyl]-4-yl)ethanone, 4-Biphenylyl methyl ketone, 4'-Phenylacetophenone, p-Phenylacetophenone, 4-Acetyl-biphenyl

A14622-22 A14622-36 A14622-14
AAA14622-22EA 115 USD
AAA14622-22 AAA14622-36 AAA14622-14
4'-Phenylacetophenone 98%
4'-Phenylacetophenone

Purity: 98% CAS No.: 92-91-1 Molecular Formula: C14H12O Formula Weight: 196.25 Melting Point: 117-123° Boiling Point: 168°/8mm MDL No.: MFCD00008749

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 168 °C/8 mm
Melting Pt: 117…123 °C
Density: 1.251 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008749
CAS Number: 92-91-1
EINECS: 202-202-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR