4'-Methoxypropiophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14619-22
A14619-36
A14619-14
AAA14619-22EA
151
USD
AAA14619-22
AAA14619-36
AAA14619-14
4'-Methoxypropiophenone 99%
4'-Methoxypropiophenone
Purity: 99%
CAS No.: 121-97-1
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 62°(143°F)
Refractive Index: 1.5465
Melting Point: 27-29°
Boiling Point: 273-275°
Beilstein Reference: 907733
MDL No.: MFCD00009310
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 273…275 °C Melting Pt: 27…29 °C Density: ~ 0.94 g/cm³ (20 °C) Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009310 CAS Number: 121-97-1 EINECS: 204-512-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...