2,3-Diaminopyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine-2,3-diyldiamine

A14618-06 A14618-22 A14618-14
AAA14618-06EA 79 USD
AAA14618-06 AAA14618-22 AAA14618-14
2,3-Diaminopyridine 98%
2,3-Diaminopyridine

Purity: 98% CAS No.: 452-58-4 Molecular Formula: C5H7N3 Formula Weight: 109.13 Flash Point: 205°(401°F) Melting Point: 110-114° Boiling Point: 195°/55mm Beilstein Reference: 109869 UN No.: 2671 MDL No.: MFCD00006319

Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 195 °C (55 mmHg)
Melting Pt: 110…114 °C
Flash Pt: 205 °C (401 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR