4-tert-Butylpyrocatechol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

A14599-0I A14599-30 A14599-0B
AAA14599-0IEA 711.63 USD
AAA14599-0I AAA14599-30 AAA14599-0B
4-tert-Butylpyrocatechol 98%
4-tert-Butylpyrocatechol

Purity: 98% CAS No.: 98-29-3 Molecular Formula: C10H14O2 Formula Weight: 166.22 Flash Point: 151°(303°F) Odor: Odorless Boiling Point: 284-286° Beilstein Reference: 2043335 UN No.: 2923 MDL No.: MFCD00002201

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 284…286 °C
Melting Pt: 53…57 °C
Density: 1.05 g/cm³ (60 °C)
Flash Pt: 151 °C (303 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR