Trimethylsulphoxonium iodide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethyloxosulphonium iodide

A14589-14 A14589-36 A14589-22
AAA14589-14EA 25.06 USD
AAA14589-14 AAA14589-36 AAA14589-22
Trimethylsulphoxonium iodide 98+%
Trimethylsulphoxonium iodide

Purity: 98+% CAS No.: 1774-47-6 Molecular Formula: (CH3)3SOI Formula Weight: 220.07 Melting Point: ca 170° dec. Beilstein Reference: 3595854

Formula: C₃H₉IOS
MW: 220.07 g/mol
Melting Pt: ∼ 170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011899
CAS Number: 1774-47-6
EINECS: 217-204-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR