2-Methyl-4-quinolinol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-2-methylquinoline

A14587-06 A14587-22 A14587-14
AAA14587-06EA 51.2 USD
AAA14587-06 AAA14587-22 AAA14587-14
2-Methyl-4-quinolinol 98+%
2-Methyl-4-quinolinol

Purity: 98+% CAS No.: 607-67-0 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 234-236° Beilstein Reference: 118320

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00006758
CAS Number: 607-67-0
EINECS: 210-140-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR