n-Tricosane 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Tricosane

A14577-06 A14577-22 A14577-14
AAA14577-06EA 51.57 USD
AAA14577-06 AAA14577-22 AAA14577-14
n-Tricosane 99%
n-Tricosane

Purity: 99% CAS No.: 638-67-5 Molecular Formula: CH3(CH2)21CH3 Formula Weight: 324.64 Flash Point: 110°(230°F) Refractive Index: 1.4468 Melting Point: 47-50° Boiling Point: 199-200°/3mm Beilstein Reference: 1756000 MDL No.: MFCD00009350

Formula: H₃C(CH₂)₂₁CH₃
MW: 324.63 g/mol
Boiling Pt: 199…200 °C/3 mm
Melting Pt: 47…50 °C
Density: 0.7969 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009350
CAS Number: 638-67-5
EINECS: 211-347-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR