1-Bromo-4-pentylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Pentylbromobenzene, 4-Bromopentylbenzene, (4-Bromophenyl)pentane
A14457-03
A14457-06
AAA14457-03EA
33
USD
AAA14457-03
AAA14457-06
1-Bromo-4-pentylbenzene 98%
1-Bromo-4-pentylbenzene
Purity: 98%
CAS No.: 51554-95-1
Molecular Formula: C11H15Br
Formula Weight: 227.15
MDL No.: MFCD00061113
Formula:
C₁₁H₁₅Br MW: 227.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061113 CAS Number: 51554-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...