Ethyl nitroacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nitroethansäureethylester, Nitroacetic ethyl ester

A14433-14 A14433-06 A14433-22
AAA14433-14EA 238 USD
AAA14433-14 AAAA14433-03 AAA14433-06 AAAA14433-22
Ethyl nitroacetate 97%
Ethyl nitroacetate

Purity: 97% CAS No.: 626-35-7 Molecular Formula: C4H7NO4 Formula Weight: 133.11 Flash Point: 92°(197°F) Refractive Index: 1.4240 Odor: Weak Boiling Point: 93-95°/10mm Beilstein Reference: 1210027 MDL No.: MFCD00007403

Formula: C₄H₇NO₄
Boiling Pt: 93…95 °C/10 mm
Density: 1.226 g/cm³ (0 °C)
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR