2,6-Dichlorophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

A14411-36 A14411-22
AAAA14411-36EA 262 USD
AAAA14411-36 AAA14411-22
2,6-Dichlorophenol 99%
2,6-Dichlorophenol

Purity: 99% CAS No.: 87-65-0 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odor: Phenol-like Melting Point: 65-68° Boiling Point: 218-220° Beilstein Reference: 1447806 Merck Reference: 14,3073 UN No.: 2020 MDL No.: MFCD00002176

Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: ~ 219 °C (1013 hPa)
Melting Pt: 68 °C
Density: 1.653 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002176
CAS Number: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1,III
Merck Index: 13,03099

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR