1,2,4,5-Tetrachlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A14318-18 A14318-09 A14318-30
AAAA14318-18EA 97.46 USD
AAAA14318-18 AAAA14318-09 AAA14318-30
1,2,4,5-Tetrachlorobenzene 98%
1,2,4,5-Tetrachlorobenzene

Purity: 98% CAS No.: 95-94-3 Molecular Formula: C6H2Cl4 Formula Weight: 215.89 Flash Point: 155°(311°F) Odor: Unpleasant Boiling Point: 245° Beilstein Reference: 1618315 UN No.: 3077 MDL No.: MFCD00000549

Formula: C₆H₂Cl₄
MW: 215.89 g/mol
Boiling Pt: 245 °C
Melting Pt: 138…142 °C
Density: 1.732 g/cm³ (20 °C)
Flash Pt: 155 °C(311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000549
CAS Number: 95-94-3
EINECS: 202-466-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR