Cyclohexyltriphenylphosphonium bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14298-14
A14298-06
AAA14298-14EA
149
USD
AAA14298-14
AAA14298-22
AAA14298-06
Cyclohexyltriphenylphosphonium bromide 98+%
Cyclohexyltriphenylphosphonium bromide
Purity: 98+%
CAS No.: 7333-51-9
Molecular Formula: C24H26BrP
Formula Weight: 425.36
Melting Point: 265-272°
Beilstein Reference: 3581012
Formula:
C₂₄H₂₆BrP MW: 425.35 Melting Pt: 265…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031667 CAS Number: 7333-51-9 EINECS: 230-834-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...