1-Octanesulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium octane-1-sulphonate

A14292-22 A14292-06 A14292-14
AAA14292-22EA 768.58 USD
AAA14292-22 AAA14292-06 AAA14292-14
1-Octanesulfonic acid sodium salt 99%
1-Octanesulphonic acid sodium salt

Reagent for HPLC analysis of peptides and proteins


Purity: 99% CAS No.: 5324-84-5 Molecular Formula: CH3(CH2)7SO3Na Formula Weight: 216.28 Melting Point: 300° Beilstein Reference: 3574241


Reagent for HPLC analysis of peptides and proteins

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: ? 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR