(4-Fluorophenyl)acetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile

A14290-14 A14290-06 A14290-22
AAA14290-14EA 60 USD
AAA14290-14 AAA14290-06 AAA14290-22
(4-Fluorophenyl)acetonitrile 98+%
(4-Fluorophenyl)acetonitrile

Purity: 98+% CAS No.: 459-22-3 Molecular Formula: C8H6FN Formula Weight: 135.14 Flash Point: 108°(226°F) Refractive Index: 1.5002 Boiling Point: 238° Beilstein Reference: 1907764 UN No.: 3276 MDL No.: MFCD00001917

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 238 °C
Melting Pt: 86 °C
Density: 1.136 g/cm³ (20 °C)
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001917
CAS Number: 459-22-3
EINECS: 207-286-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR