Broxyquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5,7-Dibromo-8-hydroxyquinoline, 5,7-Dibromo-8-quinolinol

A14284-18 A14284-30
AAA14284-18EA 53.5 USD
AAA14284-18 AAA14284-30
Broxyquinoline 98%
Broxyquinoline

Purity: 98% CAS No.: 521-74-4 Molecular Formula: C9H5Br2NO Formula Weight: 302.96 Melting Point: 198-203° Beilstein Reference: 153624 Merck Reference: 14,1453 MDL No.: MFCD00006785

Formula: C₉H₅Br₂NO
MW: 302.95 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 198…203 °C
Density: 2.189 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006785
CAS Number: 521-74-4
EINECS: 208-317-8
Merck Index: 13,01438

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR