4-Bromobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14237-14 A14237-22 A14237-36
AAA14237-14EA 89.21 USD
AAA14237-14 AAA14237-22 AAA14237-36
4-Bromobenzaldehyde 98+%
4-Bromobenzaldehyde

Purity: 98+% CAS No.: 1122-91-4 Molecular Formula: C7H5BrO Formula Weight: 185.03 Flash Point: 108°(226°F) Odor: Characteristic Boiling Point: 66-68°/2mm Beilstein Reference: 507100 MDL No.: MFCD00003377

Formula: C₇H₅BrO
Boiling Pt: 66…68 °C/2 mm
Melting Pt: 55…58 °C
Density: 1.849 g/cm³ (20 °C)
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4
EINECS: 214-365-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR