Sebacoyl dichloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sebacoyl chloride

A14226-22 A14226-14 A14226-36
AAA14226-22EA 299 USD
AAA14226-22 AAA14226-14 AAA14226-36
Sebacoyl dichloride 97%
Sebacoyl dichloride

Purity: 97% CAS No.: 111-19-3 Molecular Formula: ClCO(CH2)8COCl Formula Weight: 239.14 Flash Point: 165°(329°F) Refractive Index: 1.4680 Odor: Acrid Melting Point: -5° Boiling Point: 161°/14mm Beilstein Reference: 1365665 UN No.: 2922 MDL No.: MFCD00000770

Formula: C₁₀H₁₆Cl₂O₂
MW: 239.14 g/mol
Boiling Pt: 161 °C/14 mm
Melting Pt: –5 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR